Assay Details
| Gene Symbol: ATP6V0C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124744.4 | XP_003124792.1 | 265 | 88 | ||
| GenBank mRNA | AK231969.1 | - | 1 - 2 | 217 | 88 | |
| AK346661.1 | - | 1 - 2 | 189 | 88 | ||
| AK391769.1 | - | 1 - 2 | 243 | 88 | ||
| AY610014.1 | - | 1 - 2 | 215 | 88 | ||
More Information
Set Membership: |
|

ATP6V0C
Genomic Map