Assay Details
| Gene Symbol: CUTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356609.3 | XP_003356657.1 | 463 | 67 | ||
| XM_005665889.3 | XP_005665946.2 | 474 | 67 | |||
| GenBank mRNA | AK231998.1 | - | 3 - 4 | 676 | 67 | |
| AK348437.1 | - | 5 - 6 | 408 | 67 | ||
| AK398778.1 | - | 5 - 6 | 397 | 67 | ||
| AY610345.1 | - | 3 - 4 | 343 | 67 | ||
More Information
Set Membership: |
|

CUTA
Genomic Map