Assay Details
| Gene Symbol: NAAA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129087.4 | XP_003129135.1 | 1406 | 115 | ||
| GenBank mRNA | AK232001.1 | - | 1 - 2 | 103 | 115 | |
| AK240069.1 | - | 10 - 11 | 1363 | 115 | ||
| AK389798.1 | - | 9 - 10 | 747 | 115 | ||
More Information
Set Membership: |
|

NAAA
Genomic Map