Assay Details
| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926256.5 | XP_001926291.1 | 760 | 64 | ||
| XM_013989042.1 | XP_013844496.1 | 760 | 64 | |||
| GenBank mRNA | AK232049.1 | - | 3 - 4 | 436 | 64 | |
| AK235198.1 | - | 5 - 6 | 766 | 64 | ||
| AK395902.1 | - | 3 - 4 | 435 | 64 | ||
More Information
Set Membership: |
|

BRMS1L
Genomic Map