Assay Details
| Gene Symbol: LARP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065070.1 | XP_020920729.1 | 410 | 79 | ||
| GenBank mRNA | AK232083.1 | - | 2 - 3 | 415 | 79 | |
More Information
Set Membership: |
|

LARP4B
Genomic Map