Assay Details
| Gene Symbol: PA2G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013988304.2 | XP_013843758.1 | 1041 | 75 | ||
| XM_021091800.1 | XP_020947459.1 | 1151 | 75 | |||
| GenBank mRNA | AK232085.1 | - | 7 - 8 | 834 | 75 | |
| AK238622.1 | - | 9 - 10 | 1106 | 75 | ||
| AK346180.1 | - | 10 - 11 | 1083 | 75 | ||
| AK351877.1 | - | 9 - 10 | 975 | 75 | ||
| AK351989.1 | - | 9 - 10 | 1014 | 75 | ||
| AK352019.1 | - | 9 - 10 | 1020 | 75 | ||
| AK393697.1 | - | 9 - 10 | 1031 | 75 | ||
| AK393872.1 | - | 7 - 8 | 834 | 75 | ||
| AK396360.1 | - | 8 - 9 | 1065 | 75 | ||
| AK397997.1 | - | 9 - 10 | 973 | 75 | ||
| AK399120.1 | - | 9 - 10 | 1002 | 75 | ||
| AK399123.1 | - | 9 - 10 | 973 | 75 | ||
| AK400919.1 | - | 9 - 10 | 1005 | 75 | ||
More Information
Set Membership: |
|

PA2G4
Genomic Map