Assay Details
| Gene Symbol: ZNF608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK232091.1 | - | 1 | 1223 | 81 | |
| AK234425.1 | - | 1 | 1476 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ZNF608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK232091.1 | - | 1 | 1223 | 81 | |
| AK234425.1 | - | 1 | 1476 | 81 | ||
Set Membership: |
|