Assay Details
| Gene Symbol: BRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133367.6 | XP_003133415.3 | 675 | 96 | ||
| GenBank mRNA | AK232123.1 | - | 3 - 4 | 634 | 96 | |
| AK240416.1 | - | 3 - 4 | 630 | 96 | ||
More Information
Set Membership: |
|

BRF2
Genomic Map