Assay Details
| Gene Symbol: PDPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086751.1 | XP_020942410.1 | 924 | 60 | ||
| GenBank mRNA | AK232143.1 | - | 7 - 8 | 876 | 60 | |
| AK390136.1 | - | 7 - 8 | 881 | 60 | ||
| FJ196594.1 | - | 6 - 7 | 705 | 60 | ||
More Information
Set Membership: |
|

PDPK1
Genomic Map