Assay Details
| Gene Symbol: LOC100524359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135429.4 | XP_003135477.1 | 1231 | 87 | ||
| GenBank mRNA | AK389442.1 | - | 1 | 818 | 87 | |
More Information
Set Membership: |
|

LOC100524359
Genomic Map