Assay Details
| Gene Symbol: GALNTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134544.4 | XP_003134592.1 | 1884 | 94 | ||
| XM_013983525.2 | XP_013838979.1 | 13256 | 94 | |||
| XM_013983528.2 | XP_013838982.1 | 13237 | 94 | |||
| XM_013983534.2 | XP_013838988.1 | 12622 | 94 | |||
| XM_021078622.1 | XP_020934281.1 | 1901 | 94 | |||
| XM_021078623.1 | XP_020934282.1 | 13118 | 94 | |||
| XM_021078624.1 | XP_020934283.1 | 13033 | 94 | |||
| XM_021078625.1 | XP_020934284.1 | 1260 | 94 | |||
| XM_021078626.1 | XP_020934285.1 | 12895 | 94 | |||
| GenBank mRNA | AK389760.1 | - | 2 - 3 | 363 | 94 | |
More Information
Set Membership: |
|

GALNTL5
Genomic Map