Assay Details
| Gene Symbol: AEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126446.5 | XP_003126494.3 | 3740 | 86 | ||
| XM_021092257.1 | XP_020947916.1 | 3219 | 86 | |||
| GenBank mRNA | AK389773.1 | - | 1 | 1824 | 86 | |
More Information
Set Membership: |
|

AEBP2
Genomic Map