Assay Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076343.1 | XP_020932002.1 | 297 | 108 | ||
| GenBank mRNA | AK389869.1 | - | 2 - 3 | 200 | 108 | |
| AK399772.1 | - | 2 - 3 | 209 | 108 | ||
| AY609967.1 | - | 2 - 3 | 179 | 108 | ||
More Information
Set Membership: |
|

LEPROTL1
Genomic Map