Assay Details
| Gene Symbol: DYNLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131635.5 | XP_003131683.1 | 407 | 73 | ||
| XM_021067271.1 | XP_020922930.1 | 264 | 73 | |||
| GenBank mRNA | AK389935.1 | - | 2 - 3 | 406 | 73 | |
More Information
Set Membership: |
|

DYNLL2
Genomic Map