Assay Details
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121455.6 | XP_003121503.3 | 3912 | 100 | ||
| XM_005659487.3 | XP_005659544.1 | 1687 | 100 | |||
| XM_021093704.1 | XP_020949363.1 | 2318 | 100 | |||
| GenBank mRNA | AK390053.1 | - | 14 - 15 | 1622 | 100 | |
| AK396992.1 | - | 9 - 10 | 1173 | 100 | ||
More Information
Set Membership: |
|

NARS1
Genomic Map