Assay Details
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121455.6 | XP_003121503.3 | 2652 | 70 | ||
| XM_005659487.3 | XP_005659544.1 | 427 | 70 | |||
| XM_021093704.1 | XP_020949363.1 | 1058 | 70 | |||
| GenBank mRNA | AK390053.1 | - | 4 - 5 | 363 | 70 | |
More Information
Set Membership: |
|

NARS1
Genomic Map