Assay Details
| Gene Symbol: SYNPO2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359238.4 | XP_003359286.3 | 1219 | 87 | ||
| GenBank mRNA | AK390101.1 | - | 1 - 2 | 134 | 87 | |
More Information
Set Membership: |
|

SYNPO2L
Genomic Map