Assay Details
| Gene Symbol: PDZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927541.4 | XP_001927576.1 | 2750 | 135 | ||
| GenBank mRNA | AK390114.1 | - | 1 | 113 | 135 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PDZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927541.4 | XP_001927576.1 | 2750 | 135 | ||
| GenBank mRNA | AK390114.1 | - | 1 | 113 | 135 | |
Set Membership: |
|