Assay Details
| Gene Symbol: GABRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005666628.3 | XP_005666685.1 | 1537 | 75 | ||
| XM_013978643.2 | XP_013834097.1 | 884 | 75 | |||
| XM_013978644.2 | XP_013834098.1 | 867 | 75 | |||
| XM_013978645.2 | XP_013834099.1 | 643 | 75 | |||
| XM_021101165.1 | XP_020956824.1 | 1394 | 75 | |||
| XM_021101166.1 | XP_020956825.1 | 944 | 75 | |||
| GenBank mRNA | AK390130.1 | - | 5 - 6 | 1324 | 75 | |
| AK390188.1 | - | 4 - 5 | 642 | 75 | ||
More Information
Set Membership: |
|

GABRA2
Genomic Map