Assay Details
| Gene Symbol: ZSCAN30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127897.5 | XP_003127945.2 | 797 | 68 | ||
| GenBank mRNA | AK390131.1 | - | 3 - 4 | 773 | 68 | |
More Information
Set Membership: |
|

ZSCAN30
Genomic Map