Assay Details
Gene Symbol: LRRC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021085261.1 | XP_020940920.1 | 4112 | 97 | ||
XM_021085262.1 | XP_020940921.1 | 4904 | 97 | |||
XM_021085263.1 | XP_020940922.1 | 4294 | 97 | |||
XM_021085264.1 | XP_020940923.1 | 3568 | 97 | |||
XM_021085265.1 | XP_020940924.1 | 3918 | 97 | |||
XM_021085266.1 | XP_020940925.1 | 3487 | 97 | |||
XM_021085267.1 | XP_020940926.1 | 3922 | 97 | |||
XM_021085268.1 | XP_020940927.1 | 4057 | 97 | |||
XM_021085269.1 | XP_020940928.1 | 4545 | 97 | |||
XM_021085270.1 | XP_020940929.1 | 4275 | 97 | |||
XM_021085271.1 | XP_020940930.1 | 4190 | 97 | |||
XM_021085272.1 | XP_020940931.1 | 4194 | 97 | |||
XM_021085273.1 | XP_020940932.1 | 3484 | 97 | |||
XM_021085274.1 | XP_020940933.1 | 4109 | 97 | |||
XM_021085275.1 | XP_020940934.1 | 3403 | 97 | |||
GenBank mRNA | AK390140.1 | - | 1 | 1418 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
