Assay Details
| Gene Symbol: POU3F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926241.6 | XP_001926276.1 | 2606 | 83 | ||
| GenBank mRNA | AK390148.1 | - | 1 | 1721 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: POU3F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926241.6 | XP_001926276.1 | 2606 | 83 | ||
| GenBank mRNA | AK390148.1 | - | 1 | 1721 | 83 | |
Set Membership: |
|