Assay Details
Gene Symbol: ZNF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005670993.3 | XP_005671050.1 | 1931 | 164 | ||
GenBank mRNA | AK390160.1 | - | 1 - 2 | 189 | 164 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005670993.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
