Assay Details
| Gene Symbol: ZNF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670992.3 | XP_005671049.1 | 911 | 161 | ||
| XM_005670993.3 | XP_005671050.1 | 2268 | 161 | |||
| XM_013983246.2 | XP_013838700.1 | 2291 | 161 | |||
| XM_021072377.1 | XP_020928036.1 | 977 | 161 | |||
| XM_021072378.1 | XP_020928037.1 | 2357 | 161 | |||
| GenBank mRNA | AK390160.1 | - | 4 - 5 | 526 | 161 | |
| AK396647.1 | - | 4 - 5 | 452 | 161 | ||
More Information
Set Membership: |
|

ZNF25
Genomic Map