Assay Details
| Gene Symbol: CCDC122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013980558.2 | XP_013836012.1 | 207 | 99 | ||
| XM_013980559.2 | XP_013836013.1 | 217 | 99 | |||
| XM_013980561.2 | XP_013836015.1 | 4286 | 99 | |||
| XM_013980562.2 | XP_013836016.2 | 1187 | 99 | |||
| XM_021065619.1 | XP_020921278.1 | 1187 | 99 | |||
| XM_021065620.1 | XP_020921279.1 | 207 | 99 | |||
| XM_021065621.1 | XP_020921280.1 | 1187 | 99 | |||
| GenBank mRNA | AK390229.1 | - | 2 - 3 | 154 | 99 | |
More Information
Set Membership: |
|

CCDC122
Genomic Map