Assay Details
| Gene Symbol: ZBTB7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092801.1 | XP_020948460.1 | 222 | 79 | ||
| GenBank mRNA | AK390251.1 | - | 1 - 2 | 153 | 79 | |
| AK395365.1 | - | 1 - 2 | 157 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021092801.1 transcripts.
Set Membership: |
|

ZBTB7C
Genomic Map