Assay Details
| Gene Symbol: ZBTB7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092801.1 | XP_020948460.1 | 507 | 68 | ||
| GenBank mRNA | AK390251.1 | - | 3 - 4 | 438 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021092801.1 transcripts.
Set Membership: |
|

ZBTB7C
Genomic Map