Assay Details
| Gene Symbol: MRPS24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134862.4 | XP_003134910.2 | 160 | 91 | ||
| XM_021079350.1 | XP_020935009.1 | 168 | 91 | |||
| GenBank mRNA | AK390269.1 | - | 2 - 3 | 133 | 91 | |
| AY609730.1 | - | 2 - 3 | 121 | 91 | ||
More Information
Set Membership: |
|

MRPS24
Genomic Map