Assay Details
| Gene Symbol: ZDHHC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086470.1 | XP_020942129.1 | 2330 | 65 | ||
| XM_021086474.1 | XP_020942133.1 | 2330 | 65 | |||
| XM_021086478.1 | XP_020942137.1 | 1037 | 65 | |||
| XM_021086485.1 | XP_020942144.1 | 1042 | 65 | |||
| XM_021086493.1 | XP_020942152.1 | 2232 | 65 | |||
| XM_021086500.1 | XP_020942159.1 | 2232 | 65 | |||
| XM_021086502.1 | XP_020942161.1 | 2331 | 65 | |||
| XM_021086511.1 | XP_020942170.1 | 2622 | 65 | |||
| XM_021086517.1 | XP_020942176.1 | 2331 | 65 | |||
| XM_021086522.1 | XP_020942181.1 | 2331 | 65 | |||
| XM_021086525.1 | XP_020942184.1 | 2331 | 65 | |||
| GenBank mRNA | AK390308.1 | - | 7 - 8 | 889 | 65 | |
More Information
Set Membership: |
|

ZDHHC14
Genomic Map