Assay Details
| Gene Symbol: ONECUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093679.1 | XP_020949338.1 | 1629 | 66 | ||
| GenBank mRNA | AK390428.1 | - | 1 - 2 | 664 | 66 | |
More Information
Set Membership: |
|

ONECUT2
Genomic Map