Assay Details
| Gene Symbol: CDH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078205.1 | XP_020933864.1 | 1435 | 56 | ||
| XM_021078206.1 | XP_020933865.1 | 746 | 56 | |||
| GenBank mRNA | AK390438.1 | - | 8 - 9 | 1441 | 56 | |
More Information
Set Membership: |
|

CDH4
Genomic Map