Assay Details
| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928960.6 | XP_001928995.1 | 3088 | 138 | ||
| GenBank mRNA | AK390604.1 | - | 1 | 1515 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928960.6 | XP_001928995.1 | 3088 | 138 | ||
| GenBank mRNA | AK390604.1 | - | 1 | 1515 | 138 | |
Set Membership: |
|