Assay Details
| Gene Symbol: PRKAR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021102599.1 | XP_020958258.1 | 1347 | 63 | ||
| GenBank mRNA | AK390717.1 | - | 3 - 4 | 222 | 63 | |
More Information
Set Membership: |
|

PRKAR2B
Genomic Map