Assay Details
Gene Symbol: ACVR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927798.7 | XP_001927833.3 | 71 | |||
XM_013984146.2 | XP_013839600.1 | 71 | ||||
XM_021076448.1 | XP_020932107.1 | 71 | ||||
XM_021076449.1 | XP_020932108.1 | 71 | ||||
XM_021076450.1 | XP_020932109.1 | 71 | ||||
XM_021076451.1 | XP_020932110.1 | 71 | ||||
XM_021076452.1 | XP_020932111.1 | 71 | ||||
XM_021076453.1 | XP_020932112.1 | 71 | ||||
XM_021076454.1 | XP_020932113.1 | 71 | ||||
XM_021076455.1 | XP_020932114.1 | 71 | ||||
XM_021076456.1 | XP_020932115.1 | 71 | ||||
XM_021076457.1 | XP_020932116.1 | 71 | ||||
XM_021076458.1 | XP_020932117.1 | 71 | ||||
GenBank mRNA | AK390724.1 | - | 2 - 3 | 443 | 71 | |
HQ180176.1 | - | 3 - 4 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
