Assay Details
Gene Symbol: MKRN2OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021069346.1 | XP_020925005.1 | 643 | 68 | ||
GenBank mRNA | AK390934.1 | - | 1 - 2 | 247 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
