Assay Details
| Gene Symbol: MARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075718.1 | XP_020931377.1 | 4063 | 92 | ||
| GenBank mRNA | AK391038.1 | - | 1 | 2218 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075718.1 | XP_020931377.1 | 4063 | 92 | ||
| GenBank mRNA | AK391038.1 | - | 1 | 2218 | 92 | |
Set Membership: |
|