Assay Details
| Gene Symbol: BBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083234.1 | XP_020938893.1 | 973 | 78 | ||
| XM_021083235.1 | XP_020938894.1 | 862 | 78 | |||
| XM_021083236.1 | XP_020938895.1 | 979 | 78 | |||
| GenBank mRNA | AK391112.1 | - | 2 - 3 | 183 | 78 | |
| AK393528.1 | - | 3 - 4 | 548 | 78 | ||
More Information
Set Membership: |
|

BBOX1
Genomic Map