Assay Details
| Gene Symbol: BEND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090612.1 | XP_020946271.1 | 9433 | 77 | ||
| XM_021090618.1 | XP_020946277.1 | 8157 | 77 | |||
| GenBank mRNA | AK391217.1 | - | 1 | 1423 | 77 | |
More Information
Set Membership: |
|

BEND3
Genomic Map