Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358387.4 | XP_003358435.1 | 3720 | 64 | ||
| XM_005657067.3 | XP_005657124.1 | 3801 | 64 | |||
| XM_021071725.1 | XP_020927384.1 | 3969 | 64 | |||
| XM_021071726.1 | XP_020927385.1 | 3824 | 64 | |||
| GenBank mRNA | AK391313.1 | - | 1 | 1524 | 64 | |
More Information
Set Membership: |
|

ZNF445
Genomic Map