Assay Details
| Gene Symbol: RAP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065816.1 | XP_020921475.1 | 102 | |||
| GenBank mRNA | AK391350.1 | - | 1 | 2224 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RAP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065816.1 | XP_020921475.1 | 102 | |||
| GenBank mRNA | AK391350.1 | - | 1 | 2224 | 103 | |
Set Membership: |
|