Assay Details
| Gene Symbol: OTUD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100662.1 | XP_020956321.1 | 6884 | 117 | ||
| GenBank mRNA | AK391375.1 | - | 1 | 1915 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: OTUD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100662.1 | XP_020956321.1 | 6884 | 117 | ||
| GenBank mRNA | AK391375.1 | - | 1 | 1915 | 117 | |
Set Membership: |
|