Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925224.6 | XP_001925259.1 | 5657 | 140 | ||
| XM_003355019.4 | XP_003355067.1 | 5634 | 140 | |||
| GenBank mRNA | AK391435.1 | - | 1 | 2021 | 140 | |
More Information
Set Membership: |
|

TP53INP1
Genomic Map