Assay Details
| Gene Symbol: UGCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925267.7 | XP_001925302.2 | 3431 | 102 | ||
| XM_021067532.1 | XP_020923191.1 | 3665 | 102 | |||
| GenBank mRNA | AK391486.1 | - | 1 | 1122 | 102 | |
More Information
Set Membership: |
|

UGCG
Genomic Map