Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356445.4 | XP_003356493.1 | 1329 | 67 | ||
| XM_005665385.3 | XP_005665442.1 | 1402 | 67 | |||
| XM_013999356.2 | XP_013854810.1 | 1319 | 67 | |||
| XM_021096576.1 | XP_020952235.1 | 1392 | 67 | |||
| GenBank mRNA | AK391524.1 | - | 6 - 7 | 922 | 67 | |
More Information
Set Membership: |
|

OMA1
Genomic Map