Assay Details
| Gene Symbol: PPP4R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069219.1 | XP_020924878.1 | 1169 | 85 | ||
| XM_021069220.1 | XP_020924879.1 | 976 | 85 | |||
| GenBank mRNA | AK391551.1 | - | 1 - 2 | 227 | 85 | |
More Information
Set Membership: |
|

PPP4R2
Genomic Map