Assay Details
Gene Symbol: SH3GL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001929079.5 | XP_001929114.2 | 624 | 73 | ||
XM_013978029.2 | XP_013833483.1 | 416 | 73 | |||
XM_021098876.1 | XP_020954535.1 | 1009 | 73 | |||
GenBank mRNA | AK391635.1 | - | 5 - 6 | 536 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
