Assay Details
| Gene Symbol: ECSIT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013987373.2 | XP_013842827.1 | 915 | 92 | ||
| XM_013987374.2 | XP_013842828.1 | 740 | 92 | |||
| GenBank mRNA | AK391696.1 | - | 4 - 5 | 842 | 92 | |
| AY609803.2 | - | 4 - 5 | 821 | 92 | ||
More Information
Set Membership: |
|

ECSIT
Genomic Map