Assay Details
| Gene Symbol: DMAC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926336.6 | XP_001926371.3 | 530 | 104 | ||
| XM_013993389.2 | XP_013848843.1 | 470 | 104 | |||
| XM_021101512.1 | XP_020957171.1 | 530 | 104 | |||
| GenBank mRNA | AK391697.1 | - | 3 - 4 | 420 | 104 | |
More Information
Set Membership: |
|

DMAC2L
Genomic Map