Assay Details
| Gene Symbol: ATP5F1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122991.5 | XP_003123039.1 | 795 | 83 | ||
| GenBank mRNA | AK391792.1 | - | 1 - 2 | 210 | 83 | |
More Information
Set Membership: |
|

ATP5F1D
Genomic Map