Assay Details
| Gene Symbol: PLLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126973.5 | XP_003127021.2 | 598 | 70 | ||
| GenBank mRNA | AK391794.1 | - | 3 - 4 | 547 | 70 | |
| AK400211.1 | - | 3 - 4 | 566 | 70 | ||
More Information
Set Membership: |
|

PLLP
Genomic Map